Product Details

SNP ID
rs147619401
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:56232688 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTCACTGAATACAGGAGAGGGTT[A/G]TAGATGGCTACATAGCGATCATAAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR5T2 PubMed Links

Gene Details

Gene
OR5T2
Gene Name
olfactory receptor family 5 subfamily T member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004746.1 498 Silent Mutation TAC,TAT Y166Y NP_001004746.1

View Full Product Details