Product Details

SNP ID
rs147933718
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:56137718 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCTGGTGAAATTTTCAGGAGCCA[C/T]GTCAGAGTTTGGAAATTCATCTGTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR8J3 PubMed Links

Gene Details

Gene
OR8J3
Gene Name
olfactory receptor family 8 subfamily J member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004064.1 Intron NP_001004064.1

View Full Product Details