Product Details

SNP ID
rs148209044
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64989540 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGAGAATCATGAGGCTGTGGACC[C/T]GGAGAGAGCGGCTGAGCTGGGTAAC
Phenotype
MIM: 614983
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BATF2 PubMed Links

Gene Details

Gene
BATF2
Gene Name
basic leucine zipper ATF-like transcription factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300807.1 545 Silent Mutation CCA,CCG P114P NP_001287736.1
NM_001300808.1 545 Silent Mutation CCA,CCG P53P NP_001287737.1
NM_138456.3 545 Silent Mutation CCA,CCG P138P NP_612465.3

View Full Product Details