Product Details

SNP ID
rs148616685
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55839461 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGATGCCTTCAGCCAGTGGGCACC[A/G]CAAAGTCTTCTCCACCTGTGCCTCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR5D16 PubMed Links

Gene Details

Gene
OR5D16
Gene Name
olfactory receptor family 5 subfamily D member 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005496.1 710 Missense Mutation CAC,CGC H237R NP_001005496.1

View Full Product Details