Product Details

SNP ID
rs148622002
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5985954 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATATGTTCAGATCAATCCTTTTGT[A/G]TAAACACTCTGAGCACTCGCTGGCG
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
OR52L1 PubMed Links

Gene Details

Gene
OR52L1
Gene Name
olfactory receptor family 52 subfamily L member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005173.3 1032 Missense Mutation ACA,ATA T326I NP_001005173.3

View Full Product Details