Product Details

SNP ID
rs148659044
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:59504381 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTTCACTGCCCTCCCCACAGAAA[A/C]GGCCATCTCTGTCACCTTCACTGTC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR4D11 PubMed Links

Gene Details

Gene
OR4D11
Gene Name
olfactory receptor family 4 subfamily D member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004706.1 806 Missense Mutation AAG,ACG K269T NP_001004706.1

View Full Product Details