Product Details

SNP ID
rs148797889
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:57543205 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCAGAGGAAAGAGTGCAGCACTG[A/G]ACCCAAGGAGAAGGCTACTGATGAA
Phenotype
MIM: 613664 MIM: 603890
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SMTNL1 PubMed Links

Gene Details

Gene
SMTNL1
Gene Name
smoothelin like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105565.2 565 Missense Mutation GAA,GGA E188G NP_001099035.2
XM_005273819.3 565 Missense Mutation GAA,GGA E188G XP_005273876.1
Gene
UBE2L6
Gene Name
ubiquitin conjugating enzyme E2 L6
There are no transcripts associated with this gene.

View Full Product Details