Product Details

SNP ID
rs148853259
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:4450103 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAGGTGCCATCTTAGCCTTCTACA[C/T]AACTGTGGTCATCTCTTCAGTCATG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR52K2 PubMed Links

Gene Details

Gene
OR52K2
Gene Name
olfactory receptor family 52 subfamily K member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005172.2 809 Missense Mutation ACA,ATA T255I NP_001005172.2

View Full Product Details