Product Details

SNP ID
rs148861414
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:124310158 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTGGGCACCAAGGTGTTAAACCC[C/T]GCAATGATAAAAAGTAGAAGCTCAT
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
OR8D1 PubMed Links
Additional Information
For this assay, SNP(s) [rs4936919] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
OR8D1
Gene Name
olfactory receptor family 8 subfamily D member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002917.1 609 Silent Mutation NP_001002917.1
Gene
OR8D2
Gene Name
olfactory receptor family 8 subfamily D member 2 (gene/pseudogene)
There are no transcripts associated with this gene.

Gene
OR8G5
Gene Name
olfactory receptor family 8 subfamily G member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005198.1 609 Intron NP_001005198.1
XM_011542662.2 609 Intron XP_011540964.1
XM_011542663.2 609 Intron XP_011540965.1
XM_011542664.2 609 Intron XP_011540966.1
XM_011542665.2 609 Intron XP_011540967.1
XM_011542666.2 609 Intron XP_011540968.1
XM_011542667.2 609 Intron XP_011540969.1

View Full Product Details