Product Details

SNP ID
rs148879846
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5440891 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATGAAAGTTGAGTCCATACCATA[C/G]GTAAAAATGATCACCAAGAGCCCAT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR51B5 PubMed Links

Gene Details

Gene
OR51B5
Gene Name
olfactory receptor family 51 subfamily B member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005567.2 663 Intron NP_001005567.2
Gene
OR51I1
Gene Name
olfactory receptor family 51 subfamily I member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005288.2 663 Silent Mutation ACC,ACG T208T NP_001005288.1

View Full Product Details