Product Details

SNP ID
rs149250728
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:123906713 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGCAGAGATAGATCCATCTCCTA[G/T]TCTGGATGCATGATTCAGCTGTTTT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR8D4 PubMed Links

Gene Details

Gene
OR8D4
Gene Name
olfactory receptor family 8 subfamily D member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005197.1 282 Nonsense Mutation TAG,TAT *94Y NP_001005197.1

View Full Product Details