Product Details

SNP ID
rs149344014
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:10853306 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCCTTAGGTGCTGCACAATGGCA[C/T]TGCAAATATCTTTATCAACTGTAGA
Phenotype
MIM: 615251
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZBED5 PubMed Links

Gene Details

Gene
ZBED5
Gene Name
zinc finger BED-type containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143667.1 2139 Missense Mutation AAT,AGT N547S NP_001137139.1
NM_021211.3 2139 Missense Mutation AAT,AGT N547S NP_067034.2
Gene
ZBED5-AS1
Gene Name
ZBED5 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details