Product Details

SNP ID
rs149369852
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:73958329 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCCTCTTCCAGGTACCGCAGACT[C/T]GCCCTTAAGCACCACCCGTTGAAGT
Phenotype
MIM: 610263
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAJB13 PubMed Links

Gene Details

Gene
DNAJB13
Gene Name
DnaJ heat shock protein family (Hsp40) member B13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153614.3 1089 Silent Mutation CTC,CTT L27L NP_705842.2
XM_005273984.3 1089 Silent Mutation CTC,CTT L27L XP_005274041.1
XM_011545004.2 1089 Silent Mutation CTC,CTT L61L XP_011543306.1
XM_011545005.2 1089 Silent Mutation CTC,CTT L61L XP_011543307.1
XM_011545007.2 1089 UTR 5 XP_011543309.1
XM_011545009.2 1089 UTR 5 XP_011543311.1
XM_011545013.2 1089 Silent Mutation CTC,CTT L61L XP_011543315.1
XM_011545014.2 1089 Silent Mutation CTC,CTT L61L XP_011543316.1
XM_011545015.2 1089 Intron XP_011543317.1
XM_017017675.1 1089 UTR 5 XP_016873164.1
XM_017017676.1 1089 UTR 5 XP_016873165.1
XM_017017677.1 1089 UTR 5 XP_016873166.1
XM_017017678.1 1089 UTR 5 XP_016873167.1
XM_017017679.1 1089 Silent Mutation CTC,CTT L61L XP_016873168.1

View Full Product Details