Product Details

SNP ID
rs149446306
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:59365122 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTATCATGATATCCTATGGCTAT[A/G]TTGGCATCTCCATCATGAAGATCAC
Phenotype
MIM: 615702
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR5AN1 PubMed Links

Gene Details

Gene
OR5AN1
Gene Name
olfactory receptor family 5 subfamily AN member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004729.1 664 Missense Mutation ATT,GTT I222V NP_001004729.1

View Full Product Details