Product Details

SNP ID
rs149512166
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:58118970 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAGGGTGAAGGTGCAAGTGGTAC[A/G]CAGGATGGCTCCTGACACCCCACAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR9I1 PubMed Links

Gene Details

Gene
OR9I1
Gene Name
olfactory receptor family 9 subfamily I member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005211.1 475 Missense Mutation CGT,TGT R159C NP_001005211.1
Gene
OR9Q1
Gene Name
olfactory receptor family 9 subfamily Q member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005212.3 475 Intron NP_001005212.1

View Full Product Details