Product Details

SNP ID
rs149894937
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:47779133 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGGTCCGACGATATAATAAATCC[C/T]GTGTTGCCTTATCAACTTTTTGCTG
Phenotype
MIM: 607614
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NUP160 PubMed Links

Gene Details

Gene
NUP160
Gene Name
nucleoporin 160
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318399.1 4407 Intron NP_001305328.1
NM_015231.2 4407 Missense Mutation CAG,CGG Q1428R NP_056046.1

View Full Product Details