Product Details

SNP ID
rs149990643
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:62690838 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACCATCCTCTGTAACATCTGATT[G/T]CGGAGTTGACTCCTCCTGGCCTTCA
Phenotype
MIM: 606158
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BSCL2 PubMed Links

Gene Details

Gene
BSCL2
Gene Name
BSCL2, seipin lipid droplet biogenesis associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122955.3 1614 Missense Mutation AAA,CAA K368Q NP_001116427.1
NM_001130702.2 1614 Silent Mutation CGA,CGC R256R NP_001124174.2
NM_032667.6 1614 Missense Mutation AAA,CAA K304Q NP_116056.3
Gene
HNRNPUL2-BSCL2
Gene Name
HNRNPUL2-BSCL2 readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
LRRN4CL
Gene Name
LRRN4 C-terminal like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203422.3 1614 Intron NP_981967.1

View Full Product Details