Product Details

SNP ID
rs150319132
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:8685714 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTCTGGGAAAGGGAAAGCTCCCA[A/G]AGCAGCCTGTCATCGTGAAGGCCAA
Phenotype
MIM: 603637 MIM: 611333 MIM: 140750 MIM: 612000
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RPL27A PubMed Links

Gene Details

Gene
RPL27A
Gene Name
ribosomal protein L27a
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000990.4 721 Missense Mutation AAG,GAG K119E NP_000981.1
Gene
SNORA3A
Gene Name
small nucleolar RNA, H/ACA box 3A
There are no transcripts associated with this gene.

Gene
SNORA3B
Gene Name
small nucleolar RNA, H/ACA box 3B
There are no transcripts associated with this gene.

Gene
ST5
Gene Name
suppression of tumorigenicity 5
There are no transcripts associated with this gene.

Gene
TRIM66
Gene Name
tripartite motif containing 66
There are no transcripts associated with this gene.

View Full Product Details