Product Details

SNP ID
rs150388165
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64989572 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCGGCTGAGCTGGGTAACAGGAAC[C/T]CGGGGTCTGGAACAGCTCCAGCTGC
Phenotype
MIM: 614983
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BATF2 PubMed Links

Gene Details

Gene
BATF2
Gene Name
basic leucine zipper ATF-like transcription factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300807.1 513 Missense Mutation AGT,GGT S104G NP_001287736.1
NM_001300808.1 513 Missense Mutation AGT,GGT S43G NP_001287737.1
NM_138456.3 513 Missense Mutation AGT,GGT S128G NP_612465.3

View Full Product Details