Product Details
- SNP ID
-
rs150472827
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:108509712 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGAATTTGAGCTTAATGACTCTGAG[A/G]GGTTGGGAGGGTTCCTCAAATCATC
- Phenotype
-
MIM: 612878
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
EXPH5
PubMed Links
Gene Details
- Gene
- EXPH5
- Gene Name
- exophilin 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001144763.1 |
5906 |
Missense Mutation |
CCC,CTC |
P1856L |
NP_001138235.1 |
NM_001144764.1 |
5906 |
Missense Mutation |
CCC,CTC |
P1776L |
NP_001138236.1 |
NM_001144765.1 |
5906 |
Missense Mutation |
CCC,CTC |
P1744L |
NP_001138237.1 |
NM_001308019.1 |
5906 |
Missense Mutation |
CCC,CTC |
P1925L |
NP_001294948.1 |
NM_015065.2 |
5906 |
Missense Mutation |
CCC,CTC |
P1932L |
NP_055880.2 |
XM_011542696.2 |
5906 |
Missense Mutation |
CCC,CTC |
P1856L |
XP_011540998.1 |
XM_011542698.2 |
5906 |
Missense Mutation |
CCC,CTC |
P1856L |
XP_011541000.1 |
XM_011542700.2 |
5906 |
Missense Mutation |
CCC,CTC |
P1856L |
XP_011541002.1 |
XM_017017397.1 |
5906 |
Missense Mutation |
CCC,CTC |
P1856L |
XP_016872886.1 |
XM_017017398.1 |
5906 |
Missense Mutation |
CCC,CTC |
P1855L |
XP_016872887.1 |
XM_017017399.1 |
5906 |
Missense Mutation |
CCC,CTC |
P1856L |
XP_016872888.1 |
XM_017017400.1 |
5906 |
Missense Mutation |
CCC,CTC |
P1856L |
XP_016872889.1 |
XM_017017401.1 |
5906 |
Missense Mutation |
CCC,CTC |
P1855L |
XP_016872890.1 |
XM_017017402.1 |
5906 |
Missense Mutation |
CCC,CTC |
P1776L |
XP_016872891.1 |
XM_017017403.1 |
5906 |
Missense Mutation |
CCC,CTC |
P1775L |
XP_016872892.1 |
XM_017017404.1 |
5906 |
Missense Mutation |
CCC,CTC |
P1744L |
XP_016872893.1 |
View Full Product Details