Product Details

SNP ID
rs150544761
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:62190328 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCTGCTGCTCACGCTGGCCCTTT[G/T]CTGCTACCGGGGTGAGTACATCAGT
Phenotype
MIM: 615060
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SCGB1D1 PubMed Links

Gene Details

Gene
SCGB1D1
Gene Name
secretoglobin family 1D member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006552.1 91 Missense Mutation TGC,TTC C15F NP_006543.1

View Full Product Details