Product Details
- SNP ID
-
rs150590684
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:101915318 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGGAGGCCCGGAACCCGGAGCGCG[A/G]TCGGGGAACTGGGCACTGAATCATC
- Phenotype
-
MIM: 607666
MIM: 614634
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ANGPTL5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs117883580] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ANGPTL5
- Gene Name
- angiopoietin like 5
- Gene
- CEP126
- Gene Name
- centrosomal protein 126
View Full Product Details