Product Details

SNP ID
rs150672448
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66285531 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCACACAGGCCCAGCACCTCCGGG[C/G]AGAACCTGCGCCAAAGCAGGGGTGG
Phenotype
MIM: 611288 MIM: 612565 MIM: 611484
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CNIH2 PubMed Links

Gene Details

Gene
CNIH2
Gene Name
cornichon family AMPA receptor auxiliary protein 2
There are no transcripts associated with this gene.

Gene
RAB1B
Gene Name
RAB1B, member RAS oncogene family
There are no transcripts associated with this gene.

Gene
TMEM151A
Gene Name
transmembrane protein 151A
There are no transcripts associated with this gene.

Gene
YIF1A
Gene Name
Yip1 interacting factor homolog A, membrane trafficking protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300861.1 649 Intron NP_001287790.1
NM_020470.2 649 Missense Mutation TCC,TGC S164C NP_065203.2
XM_005273720.3 649 Missense Mutation TCC,TGC S164C XP_005273777.1
XM_017017139.1 649 Missense Mutation TCC,TGC S176C XP_016872628.1
XM_017017140.1 649 Missense Mutation TCC,TGC S176C XP_016872629.1
XM_017017141.1 649 Intron XP_016872630.1

View Full Product Details