Product Details
- SNP ID
-
rs150689610
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:118957647 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GACAGCCACTGAGTCCAGCAGAGAG[A/T]TCCCAATGTCCACACTCCCTCGTAA
- Phenotype
-
MIM: 611558
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
UPK2
PubMed Links
Gene Details
- Gene
- UPK2
- Gene Name
- uroplakin 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006760.3 |
450 |
Missense Mutation |
ATC,TTC |
I133F |
NP_006751.1 |
View Full Product Details