Product Details

SNP ID
rs150833954
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64827150 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAGATGAGCTTGGAGCGCACAAAA[C/G]GGTCCTTCAGCATCTCCCTGTGGGC
Phenotype
MIM: 613991
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CDC42BPG PubMed Links

Gene Details

Gene
CDC42BPG
Gene Name
CDC42 binding protein kinase gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017525.2 4438 Missense Mutation CCT,CGT P1430R NP_059995.2
XM_011545155.2 4438 Missense Mutation CCT,CGT P1448R XP_011543457.1
XM_011545156.2 4438 Missense Mutation CCT,CGT P1441R XP_011543458.1
XM_011545157.2 4438 Silent Mutation CCC,CCG P1415P XP_011543459.1
XM_011545158.2 4438 Silent Mutation CCC,CCG P1415P XP_011543460.1
XM_011545159.2 4438 Missense Mutation CCT,CGT P1448R XP_011543461.1
XM_011545160.2 4438 Missense Mutation CCT,CGT P1350R XP_011543462.1
XM_011545161.2 4438 Intron XP_011543463.1
XM_017017996.1 4438 Missense Mutation CCT,CGT P1447R XP_016873485.1
XM_017017997.1 4438 Intron XP_016873486.1
XM_017017998.1 4438 Intron XP_016873487.1
XM_017017999.1 4438 Missense Mutation CCT,CGT P744R XP_016873488.1

View Full Product Details