Product Details

SNP ID
rs137893701
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:104126237 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCTTCATGATCATTCATGCTGTC[C/T]TCAGTATCTAAAGAAATAAAAATAT
Phenotype
MIM: 189904
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NFYB PubMed Links

Gene Details

Gene
NFYB
Gene Name
nuclear transcription factor Y subunit beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006166.3 1028 Silent Mutation GAA,GAG E36E NP_006157.1
XM_005268908.4 1028 Silent Mutation GAA,GAG E37E XP_005268965.1
XM_005268909.4 1028 Silent Mutation GAA,GAG E37E XP_005268966.1
XM_005268910.4 1028 Silent Mutation GAA,GAG E37E XP_005268967.1

View Full Product Details