Product Details

SNP ID
rs138020482
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:67657175 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCCACGCTCACGGACAGATCCAG[G/T]TTCAACAGTTGTTTGAGGATAACAG
Phenotype
MIM: 603496
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DYRK2 PubMed Links

Gene Details

Gene
DYRK2
Gene Name
dual specificity tyrosine phosphorylation regulated kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003583.3 670 Missense Mutation GTT,TTT V17F NP_003574.1
NM_006482.2 670 Missense Mutation GTT,TTT V90F NP_006473.2
XM_017020032.1 670 Missense Mutation GTT,TTT V17F XP_016875521.1

View Full Product Details