Product Details

SNP ID
rs138740542
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:57758568 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTTATTCCTATTGCTCAGGCCTC[A/C]TCATCCATGAAGGCTCCTCTGTCTA
Phenotype
MIM: 123829 MIM: 609506 MIM: 613336 MIM: 604466
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CDK4 PubMed Links

Gene Details

Gene
CDK4
Gene Name
cyclin dependent kinase 4
There are no transcripts associated with this gene.

Gene
CYP27B1
Gene Name
cytochrome P450 family 27 subfamily B member 1
There are no transcripts associated with this gene.

Gene
MARCH9
Gene Name
membrane associated ring-CH-type finger 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138396.5 1143 Missense Mutation ATC,CTC I238L NP_612405.2
Gene
METTL1
Gene Name
methyltransferase like 1
There are no transcripts associated with this gene.

Gene
MIR6759
Gene Name
microRNA 6759
There are no transcripts associated with this gene.

View Full Product Details