Product Details

SNP ID
rs139105980
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:57235577 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGCCTGGCTTAGAAGTCTGGCC[A/G]GTCCTTCTTCAGCTTCTTATAGTCA
Phenotype
MIM: 604637 MIM: 138450 MIM: 615521
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NDUFA4L2 PubMed Links

Gene Details

Gene
NDUFA4L2
Gene Name
NDUFA4, mitochondrial complex associated like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020142.3 1129 Missense Mutation NP_064527.1
XM_005269033.2 1129 Missense Mutation XP_005269090.1
XM_011538573.2 1129 Missense Mutation XP_011536875.1
Gene
NXPH4
Gene Name
neurexophilin 4
There are no transcripts associated with this gene.

Gene
SHMT2
Gene Name
serine hydroxymethyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166356.1 1129 Intron NP_001159828.1
NM_001166357.1 1129 Intron NP_001159829.1
NM_001166358.1 1129 Intron NP_001159830.1
NM_001166359.1 1129 Intron NP_001159831.1
NM_005412.5 1129 Intron NP_005403.2
XM_011538675.2 1129 Intron XP_011536977.1
XM_011538676.2 1129 Intron XP_011536978.1
XM_011538677.2 1129 Intron XP_011536979.1
XM_011538678.2 1129 Intron XP_011536980.1
Gene
STAC3
Gene Name
SH3 and cysteine rich domain 3
There are no transcripts associated with this gene.

View Full Product Details