Product Details

SNP ID
rs139469030
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:49019138 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGGGCGCTGAGGGTGGAGGGAGA[A/G]AGGGCGCTTTAAAAAAGGGAACCAT
Phenotype
MIM: 602113 MIM: 602742
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KMT2D PubMed Links

Gene Details

Gene
KMT2D
Gene Name
lysine methyltransferase 2D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003482.3 20592 UTR 3 NP_003473.3
XM_005269162.4 20592 UTR 3 XP_005269219.1
XM_006719614.3 20592 UTR 3 XP_006719677.1
XM_006719616.3 20592 UTR 3 XP_006719679.1
XM_011538770.2 20592 UTR 3 XP_011537072.1
XM_011538771.2 20592 UTR 3 XP_011537073.1
XM_011538772.2 20592 UTR 3 XP_011537074.1
XM_011538773.2 20592 UTR 3 XP_011537075.1
XM_011538774.2 20592 UTR 3 XP_011537076.1
XM_011538776.2 20592 UTR 3 XP_011537078.1
Gene
PRKAG1
Gene Name
protein kinase AMP-activated non-catalytic subunit gamma 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206709.1 20592 Intron NP_001193638.1
NM_001206710.1 20592 Intron NP_001193639.1
NM_002733.4 20592 Intron NP_002724.1
XM_005269019.4 20592 Intron XP_005269076.1
XM_005269020.2 20592 Intron XP_005269077.1
XM_006719499.2 20592 Intron XP_006719562.1
XM_011538562.2 20592 Intron XP_011536864.1
XM_017019622.1 20592 Intron XP_016875111.1
XM_017019623.1 20592 Intron XP_016875112.1

View Full Product Details