Product Details

SNP ID
rs139622526
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:52171473 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATTTTGATCACGGGGCCTTTGCT[A/G]CCCTTCTTCTTTCGGGAGGGGGCCT
Phenotype
MIM: 611161
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRT80 PubMed Links

Gene Details

Gene
KRT80
Gene Name
keratin 80
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081492.1 1417 UTR 3 NP_001074961.1
NM_182507.2 1417 Silent Mutation GGC,GGT G428G NP_872313.2
XM_005268676.3 1417 Silent Mutation GGC,GGT G463G XP_005268733.1

View Full Product Details