Product Details

SNP ID
rs139632794
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:62603064 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTATGCCTTGGCTGCTGTTTGGCG[G/T]AGCCGGCGGCAGCCTCTGCGGGATG
Phenotype
MIM: 612148 MIM: 616822
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LINC01465 PubMed Links

Gene Details

Gene
LINC01465
Gene Name
long intergenic non-protein coding RNA 1465
There are no transcripts associated with this gene.

Gene
MIRLET7I
Gene Name
microRNA let-7i
There are no transcripts associated with this gene.

Gene
MON2
Gene Name
MON2 homolog, regulator of endosome-to-Golgi trafficking
There are no transcripts associated with this gene.

View Full Product Details