Product Details

SNP ID
rs139875282
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:49540921 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGACAGTAACTTCGTGCTGGGCAA[C/T]GCCCAGGTGGCGGGGCTCTTCCCCG
Phenotype
MIM: 604527
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KCNH3 PubMed Links

Gene Details

Gene
KCNH3
Gene Name
potassium voltage-gated channel subfamily H member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001314030.1 975 UTR 5 NP_001300959.1
NM_012284.2 975 Silent Mutation AAC,AAT N33N NP_036416.1
XM_011538085.2 975 Silent Mutation AAC,AAT N33N XP_011536387.1
XM_011538086.2 975 UTR 5 XP_011536388.1
XM_017019096.1 975 Intron XP_016874585.1
XM_017019097.1 975 Intron XP_016874586.1

View Full Product Details