Product Details

SNP ID
rs140067992
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:131930039 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGGAGAACATCCGGCGAAGAAGCT[C/G]AAGAGCGGTGGCGACGAGGAGCGGC
Phenotype
MIM: 608109 MIM: 603168
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PUS1 PubMed Links

Gene Details

Gene
PUS1
Gene Name
pseudouridylate synthase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002019.2 662 Silent Mutation CTC,CTG L41L NP_001002019.1
NM_001002020.2 662 Silent Mutation CTC,CTG L41L NP_001002020.1
NM_025215.5 662 Silent Mutation CTC,CTG L69L NP_079491.2
XM_011538768.2 662 Intron XP_011537070.1
XM_011538769.2 662 Silent Mutation CTC,CTG L69L XP_011537071.1
Gene
ULK1
Gene Name
unc-51 like autophagy activating kinase 1
There are no transcripts associated with this gene.

View Full Product Details