Product Details

SNP ID
rs140257679
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55989966 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATAGCGGCCTTCCTCACCCAGTC[C/T]GTTTGTCTAGATGACACAACAGTGA
Phenotype
MIM: 179514 MIM: 606887
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RAB5B PubMed Links

Gene Details

Gene
RAB5B
Gene Name
RAB5B, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252036.1 404 Silent Mutation TCC,TCT S61S NP_001238965.1
NM_001252037.1 404 Silent Mutation TCC,TCT S61S NP_001238966.1
NM_002868.3 404 Silent Mutation TCC,TCT S61S NP_002859.1
XM_005269051.2 404 Silent Mutation TCC,TCT S61S XP_005269108.1
XM_005269052.2 404 Silent Mutation TCC,TCT S61S XP_005269109.1
Gene
SUOX
Gene Name
sulfite oxidase
There are no transcripts associated with this gene.

View Full Product Details