Product Details

SNP ID
rs140912639
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:120324986 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGAGTATGAATAGGTGTGGGTGTA[C/T]GGGTTGTCCAGCAGAAATTTACAGC
Phenotype
MIM: 172410
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLA2G1B PubMed Links

Gene Details

Gene
PLA2G1B
Gene Name
phospholipase A2 group IB
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000928.2 306 Silent Mutation CCA,CCG P90P NP_000919.1

View Full Product Details