Product Details

SNP ID
rs142929613
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56471505 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGTGCTCATACCATGCTTTCTAA[A/G]TTCTCTTTGGACAGGGCCTCAGCTG
Phenotype
MIM: 606365
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GLS2 PubMed Links

Gene Details

Gene
GLS2
Gene Name
glutaminase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001280796.1 1957 Silent Mutation AAC,AAT N321N NP_001267725.1
NM_001280797.1 1957 Silent Mutation AAC,AAT N332N NP_001267726.1
NM_001280798.1 1957 Silent Mutation AAC,AAT N332N NP_001267727.1
NM_013267.3 1957 Silent Mutation AAC,AAT N597N NP_037399.2
XM_005268797.1 1957 Silent Mutation AAC,AAT N561N XP_005268854.1
XM_017019180.1 1957 Silent Mutation AAC,AAT N332N XP_016874669.1
Gene
SPRYD4
Gene Name
SPRY domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207344.3 1957 Intron NP_997227.1

View Full Product Details