Product Details

SNP ID
rs142954643
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:131932235 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATGACTTGGTGTCCGCCCTCGTC[A/C]GGTCAGGCTGTATTCCTGAAAATCA
Phenotype
MIM: 608109 MIM: 603168
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PUS1 PubMed Links

Gene Details

Gene
PUS1
Gene Name
pseudouridylate synthase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002019.2 819 Silent Mutation AGG,CGG R94R NP_001002019.1
NM_001002020.2 819 Silent Mutation AGG,CGG R94R NP_001002020.1
NM_025215.5 819 Silent Mutation AGG,CGG R122R NP_079491.2
XM_011538768.2 819 UTR 5 XP_011537070.1
XM_011538769.2 819 Silent Mutation AGG,CGG R122R XP_011537071.1
Gene
ULK1
Gene Name
unc-51 like autophagy activating kinase 1
There are no transcripts associated with this gene.

View Full Product Details