Product Details
- SNP ID
-
rs142967989
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:22454041 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGGAGAAAACCACTGACTCCTCCT[C/T]CCTAAATAATAGTGCACAGGAAAAT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
C2CD5
PubMed Links
Gene Details
- Gene
- C2CD5
- Gene Name
- C2 calcium dependent domain containing 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001286173.1 |
2150 |
Missense Mutation |
GAA,GGA |
E962G |
NP_001273102.1 |
NM_001286174.1 |
2150 |
Missense Mutation |
GAA,GGA |
E960G |
NP_001273103.1 |
NM_001286175.1 |
2150 |
Missense Mutation |
GAA,GGA |
E963G |
NP_001273104.1 |
NM_001286176.1 |
2150 |
Missense Mutation |
GAA,GGA |
E960G |
NP_001273105.1 |
NM_001286177.1 |
2150 |
Missense Mutation |
GAA,GGA |
E951G |
NP_001273106.1 |
NM_014802.2 |
2150 |
Missense Mutation |
GAA,GGA |
E909G |
NP_055617.1 |
XM_005253538.2 |
2150 |
Missense Mutation |
GAA,GGA |
E900G |
XP_005253595.1 |
XM_005253539.3 |
2150 |
Missense Mutation |
GAA,GGA |
E582G |
XP_005253596.1 |
XM_006719170.2 |
2150 |
Missense Mutation |
GAA,GGA |
E926G |
XP_006719233.1 |
XM_017020264.1 |
2150 |
Missense Mutation |
GAA,GGA |
E1070G |
XP_016875753.1 |
XM_017020265.1 |
2150 |
Missense Mutation |
GAA,GGA |
E1060G |
XP_016875754.1 |
XM_017020266.1 |
2150 |
Missense Mutation |
GAA,GGA |
E1053G |
XP_016875755.1 |
XM_017020267.1 |
2150 |
Missense Mutation |
GAA,GGA |
E1046G |
XP_016875756.1 |
XM_017020268.1 |
2150 |
Missense Mutation |
GAA,GGA |
E1024G |
XP_016875757.1 |
XM_017020269.1 |
2150 |
Missense Mutation |
GAA,GGA |
E1019G |
XP_016875758.1 |
XM_017020270.1 |
2150 |
Missense Mutation |
GAA,GGA |
E1006G |
XP_016875759.1 |
XM_017020271.1 |
2150 |
Missense Mutation |
GAA,GGA |
E997G |
XP_016875760.1 |
XM_017020272.1 |
2150 |
Missense Mutation |
GAA,GGA |
E973G |
XP_016875761.1 |
XM_017020273.1 |
2150 |
Missense Mutation |
GAA,GGA |
E956G |
XP_016875762.1 |
XM_017020274.1 |
2150 |
Missense Mutation |
GAA,GGA |
E950G |
XP_016875763.1 |
XM_017020275.1 |
2150 |
Missense Mutation |
GAA,GGA |
E946G |
XP_016875764.1 |
XM_017020276.1 |
2150 |
Missense Mutation |
GAA,GGA |
E941G |
XP_016875765.1 |
XM_017020277.1 |
2150 |
Missense Mutation |
GAA,GGA |
E934G |
XP_016875766.1 |
XM_017020278.1 |
2150 |
Missense Mutation |
GAA,GGA |
E922G |
XP_016875767.1 |
XM_017020279.1 |
2150 |
Missense Mutation |
GAA,GGA |
E688G |
XP_016875768.1 |
View Full Product Details