Product Details

SNP ID
rs143016679
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:119344644 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCCTTGTTAAAACAATTCAATGA[C/T]CCTCTATTGTCCTGAAAATGAAGAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC60 PubMed Links

Gene Details

Gene
CCDC60
Gene Name
coiled-coil domain containing 60
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178499.4 Intron NP_848594.2
XM_017018911.1 Intron XP_016874400.1
XM_017018912.1 Intron XP_016874401.1
XM_017018913.1 Intron XP_016874402.1
XM_017018914.1 Intron XP_016874403.1
XM_017018915.1 Intron XP_016874404.1
XM_017018916.1 Intron XP_016874405.1

View Full Product Details