Product Details

SNP ID
rs143381578
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:6845707 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACGAGAGCATCATCTGCGGCATCA[C/T]GTCCGTGGCCTTCTCCCTCAGTGGC
Phenotype
MIM: 607749 MIM: 139130 MIM: 610342 MIM: 601447
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CDCA3 PubMed Links

Gene Details

Gene
CDCA3
Gene Name
cell division cycle associated 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297602.1 1031 Intron NP_001284531.1
NM_001297603.1 1031 Intron NP_001284532.1
NM_001297604.1 1031 Intron NP_001284533.1
NM_031299.5 1031 Intron NP_112589.1
Gene
GNB3
Gene Name
G protein subunit beta 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297571.1 1031 Missense Mutation ACG,ATG T273M NP_001284500.1
NM_002075.3 1031 Missense Mutation ACG,ATG T274M NP_002066.1
XM_011520953.2 1031 Missense Mutation ACG,ATG T274M XP_011519255.1
Gene
P3H3
Gene Name
prolyl 3-hydroxylase 3
There are no transcripts associated with this gene.

Gene
USP5
Gene Name
ubiquitin specific peptidase 5
There are no transcripts associated with this gene.

View Full Product Details