Product Details

SNP ID
rs143408694
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:53443674 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTGGCTCCTGGCATGGTTGGACC[A/C]GCAGTGATAGTAGACAAGAAGATGC
Phenotype
MIM: 601210 MIM: 610459
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PCBP2 PubMed Links

Gene Details

Gene
PCBP2
Gene Name
poly(rC) binding protein 2
There are no transcripts associated with this gene.

Gene
PRR13
Gene Name
proline rich 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005354.2 Intron NP_001005354.1
NM_018457.3 Intron NP_060927.1

View Full Product Details