Product Details

SNP ID
rs144034532
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55685283 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGGTGCTTCGCCCGTTTGAAGAA[A/T]CCCATCTATAAGGACACCAGGCCAG
Phenotype
MIM: 600536
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ITGA7 PubMed Links

Gene Details

Gene
ITGA7
Gene Name
integrin subunit alpha 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144996.1 3080 Silent Mutation GGA,GGT G1067G NP_001138468.1
NM_001144997.1 3080 Silent Mutation GGA,GGT G970G NP_001138469.1
NM_002206.2 3080 Silent Mutation GGA,GGT G1063G NP_002197.2
XM_005268839.1 3080 Silent Mutation GGA,GGT G1107G XP_005268896.1
XM_005268840.1 3080 Silent Mutation GGA,GGT G1101G XP_005268897.1
XM_005268841.1 3080 Missense Mutation GAT,GTT D1145V XP_005268898.1
XM_005268842.1 3080 Silent Mutation GGA,GGT G1057G XP_005268899.1
XM_005268844.1 3080 Silent Mutation GGA,GGT G994G XP_005268901.1
XM_005268846.1 3080 Silent Mutation GGA,GGT G950G XP_005268903.1
XM_005268848.1 3080 Silent Mutation GGA,GGT G949G XP_005268905.1
XM_005268849.1 3080 Silent Mutation GGA,GGT G949G XP_005268906.1
XM_005268850.1 3080 Silent Mutation GGA,GGT G905G XP_005268907.1
XM_017019265.1 3080 Silent Mutation GGA,GGT G977G XP_016874754.1
Gene
METTL7B
Gene Name
methyltransferase like 7B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152637.2 3080 Intron NP_689850.2

View Full Product Details