Product Details

SNP ID
rs144436282
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55684963 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCCGATCCTGCCAAATCTTGATG[C/T]GACACCAGCAGCCCACTCTACCCTC
Phenotype
MIM: 600536
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ITGA7 PubMed Links

Gene Details

Gene
ITGA7
Gene Name
integrin subunit alpha 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144996.1 3400 UTR 3 NP_001138468.1
NM_001144997.1 3400 UTR 3 NP_001138469.1
NM_002206.2 3400 UTR 3 NP_002197.2
XM_005268839.1 3400 UTR 3 XP_005268896.1
XM_005268840.1 3400 UTR 3 XP_005268897.1
XM_005268841.1 3400 UTR 3 XP_005268898.1
XM_005268842.1 3400 UTR 3 XP_005268899.1
XM_005268844.1 3400 UTR 3 XP_005268901.1
XM_005268846.1 3400 UTR 3 XP_005268903.1
XM_005268848.1 3400 UTR 3 XP_005268905.1
XM_005268849.1 3400 UTR 3 XP_005268906.1
XM_005268850.1 3400 UTR 3 XP_005268907.1
XM_017019265.1 3400 UTR 3 XP_016874754.1
Gene
METTL7B
Gene Name
methyltransferase like 7B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152637.2 3400 Intron NP_689850.2

View Full Product Details