Product Details

SNP ID
rs145011914
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:2858771 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCATTGTGTCCAGGACCAGGCCT[C/T]CTGTCAGAGAACGATTGGCTGCAAG
Phenotype
MIM: 602341
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FOXM1 PubMed Links

Gene Details

Gene
FOXM1
Gene Name
forkhead box M1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243088.1 2442 Missense Mutation GAA,GGA E705G NP_001230017.1
NM_001243089.1 2442 Missense Mutation GAA,GGA E704G NP_001230018.1
NM_021953.3 2442 Missense Mutation GAA,GGA E720G NP_068772.2
NM_202002.2 2442 Missense Mutation GAA,GGA E758G NP_973731.1
NM_202003.2 2442 Missense Mutation GAA,GGA E705G NP_973732.1
XM_005253676.3 2442 Missense Mutation GAA,GGA E721G XP_005253733.1
XM_011520930.2 2442 Missense Mutation GAA,GGA E720G XP_011519232.1
XM_011520931.2 2442 Missense Mutation GAA,GGA E706G XP_011519233.1
XM_011520932.1 2442 Intron XP_011519234.1
XM_011520933.1 2442 Intron XP_011519235.1
XM_011520934.2 2442 UTR 3 XP_011519236.1
XM_011520935.1 2442 Intron XP_011519237.1
Gene
LOC100507424
Gene Name
uncharacterized LOC100507424
There are no transcripts associated with this gene.

Gene
LOC101929469
Gene Name
uncharacterized protein ENSP00000372125
There are no transcripts associated with this gene.

View Full Product Details