Product Details

SNP ID
rs146217403
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:32407771 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACACAATTTTTATCAGACTGGGGT[A/G]TCTGAGGTGAGCTCAACCGACCTGT
Phenotype
MIM: 611104
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FGD4 PubMed Links

Gene Details

Gene
FGD4
Gene Name
FYVE, RhoGEF and PH domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304480.1 Intron NP_001291409.1
NM_001304481.1 Intron NP_001291410.1
NM_001304483.1 Intron NP_001291412.1
NM_001304484.1 Intron NP_001291413.1
NM_139241.3 Intron NP_640334.2
XM_005253304.4 Intron XP_005253361.1
XM_005253307.3 Intron XP_005253364.1
XM_005253308.4 Intron XP_005253365.1
XM_005253309.1 Intron XP_005253366.1
XM_005253310.3 Intron XP_005253367.1
XM_011520554.1 Intron XP_011518856.1
XM_011520555.1 Intron XP_011518857.1
XM_011520556.1 Intron XP_011518858.1
XM_011520557.1 Intron XP_011518859.1
XM_011520558.2 Intron XP_011518860.1
XM_011520559.2 Intron XP_011518861.1
XM_017018803.1 Intron XP_016874292.1
XM_017018804.1 Intron XP_016874293.1
XM_017018805.1 Intron XP_016874294.1

View Full Product Details