Product Details

SNP ID
rs146543398
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:49333078 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAGTCGGGGTAGATGATGAAGCC[A/G]GAGAAGGTGCTGTACTTGTTGGTGT
Phenotype
MIM: 615229 MIM: 603872
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1QL4 PubMed Links

Gene Details

Gene
C1QL4
Gene Name
complement C1q like 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008223.1 1404 Silent Mutation TCC,TCT S231S NP_001008224.1
XM_011538270.2 1404 Intron XP_011536572.1
Gene
LOC101927267
Gene Name
uncharacterized LOC101927267
There are no transcripts associated with this gene.

Gene
TROAP
Gene Name
trophinin associated protein
There are no transcripts associated with this gene.

View Full Product Details