Product Details

SNP ID
rs146751372
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:53955010 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACGGGAGACACCTTCTACTTCCC[C/T]AACTTCCGCGCGTCCGGGGCGCAGC
Phenotype
MIM: 611400 MIM: 142975 MIM: 142976
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HOTAIR PubMed Links

Gene Details

Gene
HOTAIR
Gene Name
HOX transcript antisense RNA
There are no transcripts associated with this gene.

Gene
HOXC12
Gene Name
homeobox C12
There are no transcripts associated with this gene.

Gene
HOXC13
Gene Name
homeobox C13
There are no transcripts associated with this gene.

View Full Product Details