Product Details

SNP ID
rs146853631
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:109278063 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCCCAAACACGGAGTCTTTGTGA[A/G]TAACAGTTTCAACATCTTATCAGGG
Phenotype
MIM: 601557 MIM: 609429
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACACB PubMed Links

Gene Details

Gene
ACACB
Gene Name
acetyl-CoA carboxylase beta
There are no transcripts associated with this gene.

Gene
FOXN4
Gene Name
forkhead box N4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_213596.2 3456 UTR 3 NP_998761.2
XM_011537922.2 3456 UTR 3 XP_011536224.1
XM_011537923.2 3456 Intron XP_011536225.1
XM_017018818.1 3456 UTR 3 XP_016874307.1
XM_017018819.1 3456 Intron XP_016874308.1

View Full Product Details