Product Details

SNP ID
rs146968754
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55551276 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTTTATTCTACTGGGCCTTACAA[A/G]TCAACCTGAACTCCAAGTGATGATA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR6C4 PubMed Links

Gene Details

Gene
OR6C4
Gene Name
olfactory receptor family 6 subfamily C member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005494.1 50 Missense Mutation AAT,AGT N17S NP_001005494.1

View Full Product Details